Assay Details
| Gene Symbol: Sf3b6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139780.3 | NP_648037.1 | 2 - 3 | 334 | 58 | |
| GenBank mRNA | AY089603.1 | - | 2 - 3 | 341 | 58 | |
| KX531721.1 | - | 2 - 3 | 137 | 58 | ||
More Information
Set Membership: |
|

Sf3b6
Genomic Map