Assay Details
| Gene Symbol: CG10226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274544.1 | NP_001261473.1 | 7 - 8 | 3187 | 66 | |
| NM_139783.2 | NP_648040.1 | 7 - 8 | 3886 | 66 | ||
More Information
Set Membership: |
|

CG10226
Genomic Map