Assay Details
| Gene Symbol: CTCF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139852.4 | NP_648109.1 | 3 - 4 | 2487 | 72 | |
| GenBank mRNA | AY051494.1 | - | 3 - 4 | 2470 | 72 | |
More Information
Set Membership: |
|

CTCF
Genomic Map