Assay Details
| Gene Symbol: mus301 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139921.4 | NP_648178.1 | 1 - 2 | 826 | 128 | |
| GenBank mRNA | BT011090.1 | - | 1 - 2 | 820 | 128 | |
More Information
Set Membership: |
|

mus301
Genomic Map