Assay Details
| Gene Symbol: UbcE2M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274638.1 | NP_001261567.1 | 2 - 3 | 255 | 75 | |
| NM_001274639.1 | NP_001261568.1 | 2 - 3 | 382 | 75 | ||
| NM_139930.2 | NP_648187.1 | 2 - 3 | 252 | 75 | ||
| GenBank mRNA | AY118545.1 | - | 2 - 3 | 225 | 75 | |
More Information
Set Membership: |
|

UbcE2M
Genomic Map