Assay Details
| Gene Symbol: Ldh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274545.2 | NP_001261474.1 | 3 - 4 | 975 | 93 | |
| NM_001274546.1 | NP_001261475.1 | 3 - 4 | 908 | 93 | ||
| NM_001274547.1 | NP_001261476.1 | 3 - 4 | 934 | 93 | ||
| NM_057233.5 | NP_476581.1 | 3 - 4 | 975 | 93 | ||
| GenBank mRNA | BT044524.1 | - | 3 - 4 | 976 | 93 | |
| U68038.1 | - | 3 - 4 | 956 | 93 | ||
More Information
Set Membership: |
|

Ldh
Genomic Map