Assay Details
| Gene Symbol: Mybbp1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057245.3 | NP_476593.2 | 3 - 4 | 2089 | 62 | |
| GenBank mRNA | BT150288.1 | - | 3 - 4 | 2079 | 62 | |
More Information
Set Membership: |
|

Mybbp1A
Genomic Map