Assay Details
| Gene Symbol: LanA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057269.3 | NP_476617.1 | 6 - 7 | 2091 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LanA| Gene Symbol: LanA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057269.3 | NP_476617.1 | 6 - 7 | 2091 | 72 | |
Set Membership: |
|