Assay Details
| Gene Symbol: l(1)sc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057275.5 | NP_476623.1 | 1 | 847 | 136 | |
| GenBank mRNA | X71806.1 | - | 1816 | 136 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

l(1)sc| Gene Symbol: l(1)sc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057275.5 | NP_476623.1 | 1 | 847 | 136 | |
| GenBank mRNA | X71806.1 | - | 1816 | 136 | ||
Set Membership: |
|