Assay Details
| Gene Symbol: Appl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258520.2 | NP_001245449.1 | 10 - 11 | 2585 | 66 | |
| NM_001258521.2 | NP_001245450.1 | 11 - 12 | 2759 | 66 | ||
| NM_001258522.2 | NP_001245451.1 | 11 - 12 | 2744 | 66 | ||
| NM_001258523.2 | NP_001245452.1 | 11 - 12 | 2759 | 66 | ||
| NM_057278.5 | NP_476626.2 | 11 - 12 | 2756 | 66 | ||
| GenBank mRNA | AF181628.1 | - | 10 - 11 | 2731 | 66 | |
More Information
Set Membership: |
|

Appl
Genomic Map