Assay Details
| Gene Symbol: Appl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258519.1 | NP_001245448.1 | 3 - 4 | 438 | 87 | |
| NM_001258521.2 | NP_001245450.1 | 3 - 4 | 452 | 87 | ||
| NM_001258522.2 | NP_001245451.1 | 3 - 4 | 452 | 87 | ||
| NM_001258523.2 | NP_001245452.1 | 3 - 4 | 452 | 87 | ||
| NM_057278.5 | NP_476626.2 | 3 - 4 | 452 | 87 | ||
| GenBank mRNA | AF181628.1 | - | 2 - 3 | 427 | 87 | |
More Information
Set Membership: |
|

Appl
Genomic Map