Assay Details
| Gene Symbol: Appl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258519.1 | NP_001245448.1 | 4 - 5 | 584 | 69 | |
| NM_001258520.2 | NP_001245449.1 | 3 - 4 | 427 | 69 | ||
| NM_001258522.2 | NP_001245451.1 | 4 - 5 | 598 | 69 | ||
| NM_057278.5 | NP_476626.2 | 4 - 5 | 598 | 69 | ||
| GenBank mRNA | AF181628.1 | - | 3 - 4 | 573 | 69 | |
More Information
Set Membership: |
|

Appl
Genomic Map