Assay Details
| Gene Symbol: Appl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258520.2 | NP_001245449.1 | 8 - 9 | 2266 | 92 | |
| NM_001258521.2 | NP_001245450.1 | 9 - 10 | 2440 | 92 | ||
| NM_001258522.2 | NP_001245451.1 | 9 - 10 | 2425 | 92 | ||
| NM_001258523.2 | NP_001245452.1 | 9 - 10 | 2440 | 92 | ||
| NM_057278.5 | NP_476626.2 | 9 - 10 | 2437 | 92 | ||
| GenBank mRNA | AF181628.1 | - | 8 - 9 | 2412 | 92 | |
More Information
Set Membership: |
|

Appl
Genomic Map