Assay Details
| Gene Symbol: Mal-A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057279.5 | NP_476627.3 | 2 - 3 | 1544 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mal-A1| Gene Symbol: Mal-A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057279.5 | NP_476627.3 | 2 - 3 | 1544 | 96 | |
Set Membership: |
|