Assay Details
| Gene Symbol: Mal-A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057280.4 | NP_476628.2 | 1 - 2 | 191 | 68 | |
| GenBank mRNA | AY119654.1 | - | 1 - 2 | 184 | 68 | |
More Information
Set Membership: |
|

Mal-A3
Genomic Map