Assay Details
| Gene Symbol: mago | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057288.4 | NP_476636.1 | 1 - 2 | 378 | 62 | |
| GenBank mRNA | AY071060.1 | - | 1 - 2 | 376 | 62 | |
| KF051003.1 | - | 1 - 2 | 261 | 62 | ||
| KX531756.1 | - | 1 - 2 | 261 | 62 | ||
More Information
Set Membership: |
|

mago
Genomic Map