Assay Details
| Gene Symbol: Arr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259144.2 | NP_001246073.1 | 3 - 4 | 1166 | 67 | |
| NM_001299096.1 | NP_001286025.1 | 3 - 4 | 1087 | 67 | ||
| NM_057333.4 | NP_476681.1 | 3 - 4 | 1166 | 67 | ||
| GenBank mRNA | AY061824.1 | - | 2584 | 67 | ||
More Information
Set Membership: |
|

Arr1
Genomic Map