Assay Details
| Gene Symbol: Prosalpha3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057343.5 | NP_476691.1 | 2 - 3 | 297 | 82 | |
| GenBank mRNA | AY084140.1 | - | 2 - 3 | 296 | 82 | |
More Information
Set Membership: |
|

Prosalpha3
Genomic Map