Assay Details
| Gene Symbol: sgg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057368.5 | NP_476716.2 | 1 - 2 | 2331 | 77 | |
| GenBank mRNA | BT133153.1 | - | 1 - 2 | 2325 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sgg| Gene Symbol: sgg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057368.5 | NP_476716.2 | 1 - 2 | 2331 | 77 | |
| GenBank mRNA | BT133153.1 | - | 1 - 2 | 2325 | 77 | |
Set Membership: |
|