Assay Details
| Gene Symbol: alpha-Spec | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274359.1 | NP_001261288.1 | 3 - 4 | 4259 | 77 | |
| NM_057391.4 | NP_476739.1 | 3 - 4 | 4259 | 77 | ||
More Information
Set Membership: |
|

alpha-Spec
Genomic Map