Assay Details
| Gene Symbol: alpha-Spec | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274357.1 | NP_001261286.1 | 5 - 6 | 6286 | 62 | |
| NM_001274358.1 | NP_001261287.1 | 5 - 6 | 6286 | 62 | ||
| NM_001274359.1 | NP_001261288.1 | 4 - 5 | 6190 | 62 | ||
| NM_057391.4 | NP_476739.1 | 4 - 5 | 6190 | 62 | ||
More Information
Set Membership: |
|

alpha-Spec
Genomic Map