Assay Details
| Gene Symbol: Top2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273651.1 | NP_001260580.1 | 4 - 5 | 1504 | 61 | |
| NM_057412.5 | NP_476760.1 | 3 - 4 | 1249 | 61 | ||
| GenBank mRNA | BT150455.1 | - | 3 - 4 | 1244 | 61 | |
More Information
Set Membership: |
|

Top2
Genomic Map