Assay Details
| Gene Symbol: sc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057455.3 | NP_476803.1 | 1 | 1221 | 143 | |
| GenBank mRNA | BT022151.1 | - | 1 | 1196 | 143 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sc| Gene Symbol: sc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057455.3 | NP_476803.1 | 1 | 1221 | 143 | |
| GenBank mRNA | BT022151.1 | - | 1 | 1196 | 143 | |
Set Membership: |
|