Assay Details
Gene Symbol: ac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057476.3 | NP_476824.1 | 1 | 653 | 113 | |
GenBank mRNA | BT022154.1 | - | 1 | 635 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
