Assay Details
| Gene Symbol: Cbp53E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299582.1 | NP_001286511.1 | 3 - 4 | 471 | 78 | |
| NM_057490.4 | NP_476838.1 | 2 - 3 | 422 | 78 | ||
| GenBank mRNA | AY058340.1 | - | 2 - 3 | 389 | 78 | |
| X68566.1 | - | 371 | 78 | |||
More Information
Set Membership: |
|

Cbp53E
Genomic Map