Assay Details
| Gene Symbol: ex | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272894.1 | NP_001259823.1 | 3 - 4 | 963 | 77 | |
| NM_057492.4 | NP_476840.2 | 3 - 4 | 1181 | 77 | ||
| GenBank mRNA | AY069068.1 | - | 3 - 4 | 1155 | 77 | |
More Information
Set Membership: |
|

ex
Genomic Map