Assay Details
| Gene Symbol: ex | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272894.1 | NP_001259823.1 | 4 - 5 | 1032 | 80 | |
| NM_057492.4 | NP_476840.2 | 4 - 5 | 1250 | 80 | ||
| GenBank mRNA | AY069068.1 | - | 4 - 5 | 1224 | 80 | |
More Information
Set Membership: |
|

ex
Genomic Map