Assay Details
| Gene Symbol: Rrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144304.2 | NP_001137776.1 | 2 - 3 | 1329 | 58 | |
| NM_057493.4 | NP_476841.1 | 2 - 3 | 1329 | 58 | ||
| GenBank mRNA | AY118605.1 | - | 2 - 3 | 1497 | 58 | |
More Information
Set Membership: |
|

Rrp1
Genomic Map