Assay Details
| Gene Symbol: grh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299622.1 | NP_001286551.1 | 3 - 4 | 3536 | 91 | |
| NM_057494.4 | NP_476842.2 | 2 - 3 | 2074 | 91 | ||
| NM_057495.4 | NP_476843.2 | 2 - 3 | 2074 | 91 | ||
| NM_057496.5 | NP_476844.2 | 2 - 3 | 2074 | 91 | ||
| NM_057497.4 | NP_476845.2 | 2 - 3 | 2695 | 91 | ||
| GenBank mRNA | BT029431.1 | - | 2 - 3 | 1768 | 91 | |
More Information
Set Membership: |
|

grh
Genomic Map