Assay Details
| Gene Symbol: GluRIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057507.4 | NP_476855.1 | 12 - 13 | 3484 | 79 | |
| GenBank mRNA | BT050435.1 | - | 12 - 13 | 3482 | 79 | |
More Information
Set Membership: |
|

GluRIA
Genomic Map