Assay Details
| Gene Symbol: inaC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057515.3 | NP_476863.1 | 5 - 6 | 897 | 76 | |
| GenBank mRNA | BT004495.1 | - | 5 - 6 | 855 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

inaC| Gene Symbol: inaC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057515.3 | NP_476863.1 | 5 - 6 | 897 | 76 | |
| GenBank mRNA | BT004495.1 | - | 5 - 6 | 855 | 76 | |
Set Membership: |
|