Assay Details
| Gene Symbol: Su(H) | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299019.1 | NP_001285948.1 | 2 - 3 | 1714 | 92 | |
| NM_057520.4 | NP_476868.1 | 2 - 3 | 1714 | 92 | ||
| GenBank mRNA | AY069091.1 | - | 2 - 3 | 1695 | 92 | |
| M94383.1 | - | 2 - 3 | 1692 | 92 | ||
More Information
Set Membership: |
|

Su(H)
Genomic Map