Assay Details
| Gene Symbol: ple | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300026.1 | NP_001286955.1 | 6 - 7 | 1788 | 64 | |
| NM_057549.3 | NP_476897.1 | 4 - 5 | 1575 | 64 | ||
| NM_057550.4 | NP_476898.1 | 6 - 7 | 1788 | 64 | ||
| GenBank mRNA | BT044563.1 | - | 6 - 7 | 1786 | 64 | |
| X76209.1 | - | 1213 | 64 | |||
More Information
Set Membership: |
|

ple
Genomic Map