Assay Details
| Gene Symbol: ple | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300026.1 | NP_001286955.1 | 4 - 5 | 843 | 74 | |
| NM_057550.4 | NP_476898.1 | 4 - 5 | 843 | 74 | ||
| GenBank mRNA | BT044563.1 | - | 4 - 5 | 841 | 74 | |
More Information
Set Membership: |
|

ple
Genomic Map