Assay Details
| Gene Symbol: brn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057553.5 | NP_476901.1 | 1 | 1356 | 130 | |
| GenBank mRNA | BT021250.1 | - | 1 | 1354 | 130 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

brn| Gene Symbol: brn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057553.5 | NP_476901.1 | 1 | 1356 | 130 | |
| GenBank mRNA | BT021250.1 | - | 1 | 1354 | 130 | |
Set Membership: |
|