Assay Details
| Gene Symbol: Pabp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057554.4 | NP_476902.1 | 4 - 5 | 1040 | 82 | |
| NM_165589.3 | NP_724648.1 | 4 - 5 | 1040 | 82 | ||
| GenBank mRNA | AY070631.1 | - | 4 - 5 | 1038 | 82 | |
| AY075519.1 | - | 4 - 5 | 1038 | 82 | ||
More Information
Set Membership: |
|

Pabp2
Genomic Map