Assay Details
| Gene Symbol: gammaTub37C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273636.1 | NP_001260565.1 | 3 - 4 | 917 | 61 | |
| NM_057574.5 | NP_476922.1 | 3 - 4 | 917 | 61 | ||
| GenBank mRNA | AY070558.1 | - | 3 - 4 | 890 | 61 | |
More Information
Set Membership: |
|

gammaTub37C
Genomic Map