Assay Details
| Gene Symbol: Rbp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014462.3 | NP_001014462.1 | 4 - 5 | 1049 | 76 | |
| NM_001273045.1 | NP_001259974.1 | 4 - 5 | 1046 | 76 | ||
| NM_001298640.1 | NP_001285569.1 | 2 - 3 | 828 | 76 | ||
| NM_057588.3 | NP_476936.3 | 3 - 4 | 1134 | 76 | ||
| NM_057589.3 | NP_476937.3 | 3 - 4 | 1134 | 76 | ||
| NM_134297.5 | NP_599124.1 | 4 - 5 | 1050 | 76 | ||
| NM_134298.5 | NP_599125.1 | 4 - 5 | 1051 | 76 | ||
| NM_134299.5 | NP_599126.1 | 4 - 5 | 1063 | 76 | ||
| NM_134300.4 | NP_599127.1 | 4 - 5 | 1066 | 76 | ||
| GenBank mRNA | BT011197.1 | - | 4 - 5 | 1024 | 76 | |
More Information
Set Membership: |
|

Rbp9
Genomic Map