Assay Details
| Gene Symbol: Rbp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014462.3 | NP_001014462.1 | 5 - 6 | 1308 | 92 | |
| NM_001273045.1 | NP_001259974.1 | 5 - 6 | 1305 | 92 | ||
| NM_057588.3 | NP_476936.3 | 4 - 5 | 1393 | 92 | ||
| NM_134298.5 | NP_599125.1 | 5 - 6 | 1310 | 92 | ||
| NM_134299.5 | NP_599126.1 | 5 - 6 | 1322 | 92 | ||
| GenBank mRNA | BT011197.1 | - | 5 - 6 | 1283 | 92 | |
More Information
Set Membership: |
|

Rbp9
Genomic Map