Assay Details
| Gene Symbol: Rbp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014462.3 | NP_001014462.1 | 7 - 8 | 1771 | 62 | |
| NM_001273045.1 | NP_001259974.1 | 7 - 8 | 1768 | 62 | ||
| NM_001298640.1 | NP_001285569.1 | 6 - 7 | 1565 | 62 | ||
| NM_057588.3 | NP_476936.3 | 6 - 7 | 1856 | 62 | ||
| NM_057589.3 | NP_476937.3 | 7 - 8 | 1871 | 62 | ||
| NM_134297.5 | NP_599124.1 | 8 - 9 | 1787 | 62 | ||
| NM_134298.5 | NP_599125.1 | 7 - 8 | 1773 | 62 | ||
| NM_134299.5 | NP_599126.1 | 7 - 8 | 1785 | 62 | ||
| NM_134300.4 | NP_599127.1 | 8 - 9 | 1803 | 62 | ||
| GenBank mRNA | BT011197.1 | - | 7 - 8 | 1746 | 62 | |
More Information
Set Membership: |
|

Rbp9
Genomic Map