Assay Details
| Gene Symbol: Trf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057591.4 | NP_476939.1 | 1 - 2 | 231 | 67 | |
| GenBank mRNA | AY128483.1 | - | 1 - 2 | 229 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trf| Gene Symbol: Trf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057591.4 | NP_476939.1 | 1 - 2 | 231 | 67 | |
| GenBank mRNA | AY128483.1 | - | 1 - 2 | 229 | 67 | |
Set Membership: |
|