Assay Details
| Gene Symbol: Cks30A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057599.4 | NP_476947.1 | 1 - 2 | 352 | 81 | |
| GenBank mRNA | AY061285.1 | - | 1 - 2 | 333 | 81 | |
| KX531758.1 | - | 1 - 2 | 60 | 81 | ||
| U40077.1 | - | 1 - 2 | 81 | 81 | ||
More Information
Set Membership: |
|

Cks30A
Genomic Map