Assay Details
| Gene Symbol: ox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057637.5 | NP_476985.1 | 1 - 2 | 233 | 65 | |
| GenBank mRNA | KX531584.1 | - | 1 - 2 | 133 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ox| Gene Symbol: ox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057637.5 | NP_476985.1 | 1 - 2 | 233 | 65 | |
| GenBank mRNA | KX531584.1 | - | 1 - 2 | 133 | 65 | |
Set Membership: |
|