Assay Details
| Gene Symbol: RyR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259277.1 | NP_001246206.1 | 10 - 11 | 3681 | 65 | |
| NM_001259279.2 | NP_001246208.1 | 10 - 11 | 3646 | 65 | ||
| NM_001259280.1 | NP_001246209.1 | 10 - 11 | 3902 | 65 | ||
| NM_001259281.2 | NP_001246210.1 | 11 - 12 | 3879 | 65 | ||
| NM_057643.4 | NP_476991.1 | 10 - 11 | 3646 | 65 | ||
| NM_057644.4 | NP_476992.1 | 10 - 11 | 3646 | 65 | ||
More Information
Set Membership: |
|

RyR
Genomic Map