Assay Details
| Gene Symbol: RyR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259280.1 | NP_001246209.1 | 13 - 14 | 6025 | 81 | |
| NM_001259281.2 | NP_001246210.1 | 14 - 15 | 6002 | 81 | ||
| NM_001259282.2 | NP_001246211.1 | 12 - 13 | 5807 | 81 | ||
| NM_057643.4 | NP_476991.1 | 13 - 14 | 5769 | 81 | ||
| NM_057645.3 | NP_476993.1 | 13 - 14 | 5804 | 81 | ||
More Information
Set Membership: |
|

RyR
Genomic Map