Assay Details
| Gene Symbol: RyR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259277.1 | NP_001246206.1 | 15 - 16 | 6077 | 106 | |
| NM_001259278.2 | NP_001246207.1 | 16 - 17 | 6275 | 106 | ||
| NM_001259279.2 | NP_001246208.1 | 15 - 16 | 6060 | 106 | ||
| NM_001259280.1 | NP_001246209.1 | 15 - 16 | 6340 | 106 | ||
| NM_001259281.2 | NP_001246210.1 | 16 - 17 | 6317 | 106 | ||
| NM_001259282.2 | NP_001246211.1 | 14 - 15 | 6122 | 106 | ||
| NM_057643.4 | NP_476991.1 | 15 - 16 | 6084 | 106 | ||
| NM_057644.4 | NP_476992.1 | 15 - 16 | 6042 | 106 | ||
| NM_057645.3 | NP_476993.1 | 15 - 16 | 6119 | 106 | ||
| NM_057646.4 | NP_476994.1 | 15 - 16 | 6042 | 106 | ||
More Information
Set Membership: |
|

RyR
Genomic Map