Assay Details
| Gene Symbol: RyR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259277.1 | NP_001246206.1 | 27 - 28 | 12853 | 113 | |
| NM_001259278.2 | NP_001246207.1 | 27 - 28 | 13045 | 113 | ||
| NM_001259279.2 | NP_001246208.1 | 26 - 27 | 12830 | 113 | ||
| NM_001259280.1 | NP_001246209.1 | 26 - 27 | 13098 | 113 | ||
| NM_001259281.2 | NP_001246210.1 | 27 - 28 | 13084 | 113 | ||
| NM_001259282.2 | NP_001246211.1 | 25 - 26 | 12913 | 113 | ||
| NM_057643.4 | NP_476991.1 | 26 - 27 | 12854 | 113 | ||
| NM_057644.4 | NP_476992.1 | 26 - 27 | 12812 | 113 | ||
| NM_057645.3 | NP_476993.1 | 26 - 27 | 12889 | 113 | ||
| NM_057646.4 | NP_476994.1 | 26 - 27 | 12812 | 113 | ||
More Information
Set Membership: |
|

RyR
Genomic Map