Assay Details
| Gene Symbol: Mef2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382029.1 | NP_001369071.1 | 4 - 5 | 1634 | 66 | |
| NM_057670.5 | NP_477018.1 | 4 - 5 | 1634 | 66 | ||
| NM_057672.5 | NP_477020.1 | 5 - 6 | 2017 | 66 | ||
| GenBank mRNA | BT133393.1 | - | 4 - 5 | 1611 | 66 | |
More Information
Set Membership: |
|

Mef2
Genomic Map