Assay Details
| Gene Symbol: Mef2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273913.2 | NP_001260842.1 | 6 - 7 | 1792 | 73 | |
| NM_001382027.1 | NP_001369069.1 | 5 - 6 | 1714 | 73 | ||
| NM_001382029.1 | NP_001369071.1 | 5 - 6 | 1732 | 73 | ||
| NM_057670.5 | NP_477018.1 | 5 - 6 | 1732 | 73 | ||
| NM_057671.5 | NP_477019.1 | 5 - 6 | 1714 | 73 | ||
| NM_057672.5 | NP_477020.1 | 6 - 7 | 2115 | 73 | ||
| NM_057673.5 | NP_477021.1 | 5 - 6 | 1714 | 73 | ||
| GenBank mRNA | BT133393.1 | - | 5 - 6 | 1709 | 73 | |
| X83527.1 | - | 1696 | 73 | |||
More Information
Set Membership: |
|

Mef2
Genomic Map