Assay Details
| Gene Symbol: pav | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057677.4 | NP_477025.1 | 5 - 6 | 2340 | 77 | |
| GenBank mRNA | BT053676.1 | - | 5 - 6 | 2338 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pav| Gene Symbol: pav | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057677.4 | NP_477025.1 | 5 - 6 | 2340 | 77 | |
| GenBank mRNA | BT053676.1 | - | 5 - 6 | 2338 | 77 | |
Set Membership: |
|